Learn More
Waters Corp Waters Corporation Styragel HR 0.5 Column, 5 μm, 50 Å, 4.6 mm × 300 mm
Catalog No. 50785585 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • High-resolution GPC column for characterizing samples in the low to mid-molecular range.
  • Features 5 μm spherical polymer particles and a 50 Å pore size.
  • Narrow bore design reduces solvent consumption and disposal costs.
  • Operates within a pH range of 2 to 12.
  • Supports a molecular weight range from 0 to 1000.
  • Dimensions are 4.6 mm inner diameter × 300 mm length.
  • Packed with Toluene for HPLC systems.
Catalog No. 50-785-585 Supplier Waters Corp Supplier No. WAT045830
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • High-resolution GPC column for characterizing samples in the low to mid-molecular range.
  • Features 5 μm spherical polymer particles and a 50 Å pore size.
  • Narrow bore design reduces solvent consumption and disposal costs.
  • Operates within a pH range of 2 to 12.
  • Supports a molecular weight range from 0 to 1000.
  • Dimensions are 4.6 mm inner diameter × 300 mm length.
  • Packed with Toluene for HPLC systems.