Learn More
Waters Corp Styragel Column, 5 μm, 50 Å, 7.8 mm X 300 mm
Catalog No. 50785552 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HR columns are high-resolution GPC columns packed with 5 μm particles. Ideal for characterizing samples in the low to mid molecular range, they analyze low-molecular-weight polymers, oligomers, and additives with high plate counts. These columns are made of high-performance, totally porous, and extremely cross-linked styrene-divinylbenzene copolymer particles.
  • High-resolution GPC columns
  • Packed with 5 μm particles
  • Ideal for characterizing samples in the low to mid molecular range, including low-molecular-weight polymers, oligomers, and additives
  • High plate counts
  • Made from high-performance, totally porous, and extremely cross-linked styrene-divinylbenzene copolymer particles
  • Delivers consistent, dependable results
  • Manufactured to the highest industry standards for ISO, FDA, and cGMP criteria
Catalog No. 50-785-552 Supplier Waters Corp Supplier No. WAT044230
Add to Cart
Add to Cart
Styragel HR columns are high-resolution GPC columns packed with 5 μm particles. Ideal for characterizing samples in the low to mid molecular range, they analyze low-molecular-weight polymers, oligomers, and additives with high plate counts. These columns are made of high-performance, totally porous, and extremely cross-linked styrene-divinylbenzene copolymer particles.
  • High-resolution GPC columns
  • Packed with 5 μm particles
  • Ideal for characterizing samples in the low to mid molecular range, including low-molecular-weight polymers, oligomers, and additives
  • High plate counts
  • Made from high-performance, totally porous, and extremely cross-linked styrene-divinylbenzene copolymer particles
  • Delivers consistent, dependable results
  • Manufactured to the highest industry standards for ISO, FDA, and cGMP criteria