Learn More
Waters Corp Waters Styragel Column, 5 μm, 7.8 mm x 300 mm, 50 Å Pore
Supplier:  Waters Corp WAT044231
Encompass_Preferred
Styragel HR columns are high-resolution GPC columns designed for the characterization of samples in the low to mid-molecular weight range. They are packed with high-performance, fully porous, highly cross-linked styrene-divinylbenzene copolymer particles, ensuring high efficiency and unparalleled resolution.
  • Packed with 5 μm particles for high plate counts.
  • Ideal for analyzing low-molecular-weight polymers, oligomers, and additives.
  • Manufactured under strict conditions and protocols adhering to ISO, FDA, and cGMP guidelines.
  • Delivers reliable results, reducing the need for repeated analysis.
  • Features a pH range of 2 to 12.
Catalog No. 50-785-553