Learn More
Waters Corp Waters Styragel Column, 5 μm, 7.8 mm x 300 mm, 50 Å
Catalog No. 50785554 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These high-resolution GPC columns, packed with 5 μm particles, are ideal for characterizing samples in the low to mid molecular weight range, ensuring high efficiency and dependable results.
  • Offers unmatched resolution for low to mid molecular weight range samples.
  • Loaded with high-performance, totally porous, strongly cross-linked styrene-divinylbenzene copolymer particles.
  • Provides high plate counts for analysis of additives, oligomers, and low molecular weight polymers.
  • Manufactured adhering to stringent ISO, FDA, and cGMP regulations.
  • Features a broad operating pH range of 2 to 12 pH.
Catalog No. 50-785-554 Supplier Waters Corp Supplier No. WAT044232
These high-resolution GPC columns, packed with 5 μm particles, are ideal for characterizing samples in the low to mid molecular weight range, ensuring high efficiency and dependable results.
  • Offers unmatched resolution for low to mid molecular weight range samples.
  • Loaded with high-performance, totally porous, strongly cross-linked styrene-divinylbenzene copolymer particles.
  • Provides high plate counts for analysis of additives, oligomers, and low molecular weight polymers.
  • Manufactured adhering to stringent ISO, FDA, and cGMP regulations.
  • Features a broad operating pH range of 2 to 12 pH.