Learn More
Waters Corp Waters Styragel HR 1 Column, 5 μm Particle, 100 Å Pore, 4.6 x 300 mm
Catalog No. 50785588 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These narrow bore columns are designed to reduce solvent consumption and disposal costs significantly. They are high-resolution GPC columns, packed with 5 μm particles, making them ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • Reduces solvent consumption and disposal costs by up to two-thirds.
  • High-resolution GPC columns.
  • Ideal for characterizing samples in the low- to mid-molecular range.
  • Packed with 5 μm particles.
Catalog No. 50-785-588 Supplier Waters Corp Supplier No. WAT045845
These narrow bore columns are designed to reduce solvent consumption and disposal costs significantly. They are high-resolution GPC columns, packed with 5 μm particles, making them ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • Reduces solvent consumption and disposal costs by up to two-thirds.
  • High-resolution GPC columns.
  • Ideal for characterizing samples in the low- to mid-molecular range.
  • Packed with 5 μm particles.