Learn More
Waters Corp Waters GPC Column, 5 μm, 4.6 mm x 300 mm, 100 Å
Catalog No. 50785589 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This GPC column is a high-resolution tool ideal for characterizing samples in the low- to mid-molecular range. Its narrow bore design significantly reduces solvent consumption and disposal costs. Packed with 5 μm particles, it provides exceptional resolution and efficiency for accurate chromatographic studies.
  • Narrow bore design can cut solvent consumption and disposal costs by up to two-thirds.
  • Ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • Packed with hard 5 μm particles to achieve unequaled resolution and efficiency.
  • Ensures reliable results without the need to repeat analysis.
  • Manufactured under stringent conditions, adhering to ISO, FDA, and cGMP criteria, to ensure high quality.
  • Suitable for analysis of oligomers, epoxies, and polymer additives.
Catalog No. 50-785-589 Supplier Waters Corp Supplier No. WAT045850
This GPC column is a high-resolution tool ideal for characterizing samples in the low- to mid-molecular range. Its narrow bore design significantly reduces solvent consumption and disposal costs. Packed with 5 μm particles, it provides exceptional resolution and efficiency for accurate chromatographic studies.
  • Narrow bore design can cut solvent consumption and disposal costs by up to two-thirds.
  • Ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • Packed with hard 5 μm particles to achieve unequaled resolution and efficiency.
  • Ensures reliable results without the need to repeat analysis.
  • Manufactured under stringent conditions, adhering to ISO, FDA, and cGMP criteria, to ensure high quality.
  • Suitable for analysis of oligomers, epoxies, and polymer additives.