Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Styragel HR 1 Column, 5 μm, 100 Å, 7.8 mm x 300 mm

Catalog No. 50785555 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • Particle size: 5 μm
  • Pore size: 100 Å
  • Internal diameter: 7.8 mm
  • Length: 300 mm
  • Particle substrate: Polymer
  • Particle shape: Spherical
  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • pH range: 2 to 12 pH
  • Molecular weight range: 100 to 5000
  • Endfitting type: Waters
  • Packing solvent: Toluene
  • System: HPLC
  • Units per package: 1 pk

Catalog No. 50-785-555 Supplier Waters Corp Supplier No. WAT044233
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • Particle size: 5 μm
  • Pore size: 100 Å
  • Internal diameter: 7.8 mm
  • Length: 300 mm
  • Particle substrate: Polymer
  • Particle shape: Spherical
  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • pH range: 2 to 12 pH
  • Molecular weight range: 100 to 5000
  • Endfitting type: Waters
  • Packing solvent: Toluene
  • System: HPLC
  • Units per package: 1 pk

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.