Learn More
Waters Corp Waters Styragel HR Column, 5 μm, 100 Å, 7.8 mm x 300 mm
Catalog No. 50785557 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
The Styragel HR 1 column is a high-resolution GPC column ideal for characterizing low- to mid-molecular weight samples. It features 5 μm styrene-divinylbenzene copolymer particles with a 100 Å pore size and dimensions of 7.8 mm x 300 mm. This column delivers high plate counts for precise analysis of polymers, epoxies, oligomers, and additives.
  • High-resolution GPC columns.
  • Packed with 5 μm particles for superior performance.
  • Ideal for low- to mid-molecular weight sample characterization.
  • Provides high plate counts for precise analysis of polymers, epoxies, oligomers, and additives.
  • Manufactured under stringent ISO, FDA, and cGMP guidelines.
  • Ensures high quality, consistency, and reproducibility for liquid chromatography.
Catalog No. 50-785-557 Supplier Waters Corp Supplier No. WAT044235
Add to Cart
Add to Cart
The Styragel HR 1 column is a high-resolution GPC column ideal for characterizing low- to mid-molecular weight samples. It features 5 μm styrene-divinylbenzene copolymer particles with a 100 Å pore size and dimensions of 7.8 mm x 300 mm. This column delivers high plate counts for precise analysis of polymers, epoxies, oligomers, and additives.
  • High-resolution GPC columns.
  • Packed with 5 μm particles for superior performance.
  • Ideal for low- to mid-molecular weight sample characterization.
  • Provides high plate counts for precise analysis of polymers, epoxies, oligomers, and additives.
  • Manufactured under stringent ISO, FDA, and cGMP guidelines.
  • Ensures high quality, consistency, and reproducibility for liquid chromatography.