Learn More
Waters Corp Waters Styragel HR 1, 5 μm, 100 Å, 7.8 mm ID × 300 mm
Supplier: Waters Corp WAT044234
Styragel HR columns are high-resolution GPC columns packed with 5 μm particles. They are designed for characterizing samples in the low to mid molecular range, providing high resolution and efficiency for analyzing low molecular weight samples such as oligomers, epoxies, and polymer additives.
- High-resolution GPC column for critical applications.
- Packed with 5 μm rigid particles for high efficiency.
- Ideal for characterizing samples in the low to mid molecular weight region.
- Suitable for low molecular weight samples, including oligomers, epoxies, and polymer additives.
- Molecular weight range of 100 to 5,000.
- pH range of 2 to 12.
- Inner diameter of 7.8 mm.
- Length of 300 mm.
- One column included per package.
- Shipped in toluene, tetrahydrofuran (THF), and dimethylformamide (DMF).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.