Learn More
Waters Corp Styragel Column, HR 2, 5 µm, 500Å, 4.6 mm X 300 mm, 500 - 20K, DMF, 1/pk
Supplier:  Waters Corp WAT045870
Encompass_Preferred
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
Catalog No. 50-785-593