Learn More
Waters Corp Waters Column, 500 Å, 5 μm, 4.6 mm x 300 mm
Supplier: Waters Corp WAT045860
These narrow bore columns can cut solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
- Separation mode: SEC/GPC
- Particle substrate: Polymer
- pH range: 2 to 12 pH
- Molecular weight range: 500 to 20000
- Particle shape: Spherical
- Particle size: 5 μm
- Pore size: 500 Å
- Packing solvent: Toluene
- System: HPLC
- Inner diameter: 4.6 mm
- Length: 300 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.