Learn More
Waters Corp Waters Column, 1000 Å, 5 µm, 4.6 mm, 300 mm
Catalog No. 50785595 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • Narrow bore design reduces solvent consumption and disposal costs by as much as two-thirds.
  • High-resolution GPC columns.
  • Packed with 5 µm particles.
  • Ideal for characterizing samples in the low- to mid-molecular range.
  • Provides critical high resolution.
Catalog No. 50-785-595 Supplier Waters Corp Supplier No. WAT045880
Add to Cart
Add to Cart
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • Narrow bore design reduces solvent consumption and disposal costs by as much as two-thirds.
  • High-resolution GPC columns.
  • Packed with 5 µm particles.
  • Ideal for characterizing samples in the low- to mid-molecular range.
  • Provides critical high resolution.