Learn More
Waters Corp Waters Styragel Column, 1000 Å, 5 μm, 4.6 mm x 300 mm, 500 to 30000
Supplier: Waters Corp WAT045885
These narrow bore columns are high-resolution GPC columns packed with 5 μm particles. They are designed for characterizing samples in the low- to mid-molecular range where high resolution is critical, and can reduce solvent consumption and disposal costs by up to two-thirds.
- GPC chemistry for characterization.
- SEC/GPC separation mode.
- Polymer particle substrate.
- pH range of 2 to 12.
- Molecular weight range of 500 to 30000.
- Spherical particle shape.
- Particle size of 5 μm.
- Endfitting type.
- Pore size of 1000 Å.
- Column format.
- Packing solvent is DMF.
- Designed for HPLC systems.
- Inner diameter of 4.6 mm.
- Length of 300 mm.
- 1 unit per package.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.