Learn More
Waters Corp Waters Styragel HR 3 Column, 5 μm, 7.8 mm × 300 mm, 500 to 30K MW, THF
Supplier:  Waters Corp WAT044222
Encompass_Preferred
Styragel HR columns are high-resolution GPC columns packed with 5 μm particles, ideal for characterizing samples in the low to mid molecular range where high resolution is critical.
  • Packed with rigid 5 μm particles, allowing for unrivaled resolution and efficiency in the low-to-mid molecular weight region.
  • Provides critical information that is difficult to obtain by other methods.
  • Suitable for analysis of oligomers, epoxies, and polymer additives, particularly for low molecular weight samples where high resolution is critical.
  • Can be used for expanding size-exclusion techniques beyond original polymer analysis to separate small and large molecules from interfering matrices found in foods, pharmaceutical preparations, and natural products.
Catalog No. 50-785-544