Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Styragel HR 3 Column, 5 μm Particle, 7.8 mm ID, 300 mm Length, Toluene

Catalog No. 50785543 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • GPC chemistry
  • SEC/GPC separation mode
  • Polymer particle substrate
  • PH range of 2 to 12
  • Molecular weight range of 500 to 30,000
  • Spherical particle shape
  • 5 μm particle size
  • 1000 Å pore size
  • Column format
  • Toluene packing solvent
  • HPLC system compatibility
  • 7.8 mm inner diameter
  • 300 mm length
  • 1 unit per package
  • Contains hazardous material requiring special freight handling

Catalog No. 50-785-543 Supplier Waters Corp Supplier No. WAT044221
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • GPC chemistry
  • SEC/GPC separation mode
  • Polymer particle substrate
  • PH range of 2 to 12
  • Molecular weight range of 500 to 30,000
  • Spherical particle shape
  • 5 μm particle size
  • 1000 Å pore size
  • Column format
  • Toluene packing solvent
  • HPLC system compatibility
  • 7.8 mm inner diameter
  • 300 mm length
  • 1 unit per package
  • Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.