Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Corporation Column, Styragel HR 3, 1000 Å, 5 μm, 7.8 mm x 300 mm

Catalog No. 50785545 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • High-resolution GPC columns
  • Packed with 5 μm particles
  • Ideal for characterizing samples in the low to mid molecular range
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Molecular weight range: 500 to 30000
  • Particle shape: Spherical
  • Pore size: 1000 Å
  • Packing solvent: DMF
  • System: HPLC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • 1 unit per package

Catalog No. 50-785-545 Supplier Waters Corp Supplier No. WAT044223
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • High-resolution GPC columns
  • Packed with 5 μm particles
  • Ideal for characterizing samples in the low to mid molecular range
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12 pH
  • Molecular weight range: 500 to 30000
  • Particle shape: Spherical
  • Pore size: 1000 Å
  • Packing solvent: DMF
  • System: HPLC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • 1 unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.