Learn More
Waters Corp Waters Corporation Column, 5 μm, 10000 Å, 4.6 mm x 300 mm
Catalog No. 50785599 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These narrow bore columns can cut solvent consumption and disposal costs by as much as two-thirds. They are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • Can cut solvent consumption and disposal costs by as much as two-thirds.
  • High-resolution GPC columns.
  • Ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
Catalog No. 50-785-599 Supplier Waters Corp Supplier No. WAT045900
These narrow bore columns can cut solvent consumption and disposal costs by as much as two-thirds. They are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
  • Can cut solvent consumption and disposal costs by as much as two-thirds.
  • High-resolution GPC columns.
  • Ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.