Learn More
Waters Corp Styragel Column, HR 4, 5 µm, 10-4Å, 4.6 mm X 300 mm, 5K - 600K, THF, 1/pk
Catalog No. 50785598 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
Catalog No. 50-785-598 Supplier Waters Corp Supplier No. WAT045895
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.