Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Styragel HR 4 Column, 5 μm, 4.6 mm x 300 mm

Catalog No. 50785597 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These narrow bore columns significantly reduce solvent consumption and disposal costs. As high-resolution GPC columns with 5 μm particles, they are ideal for characterizing low- to mid-molecular weight samples requiring high resolution.

  • High-resolution GPC column
  • Packed with 5 μm spherical particles
  • Ideal for low- to mid-molecular weight range samples
  • Narrow bore design reduces solvent and disposal costs
  • Compatible with HPLC systems
  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range: 2 to 12
  • Molecular weight range: 5000 to 600000
  • Pore size: 10000 Å
  • Packed in toluene
  • Dimensions: 4.6 mm inner diameter x 300 mm length
  • Single unit per package

Catalog No. 50-785-597 Supplier Waters Corp Supplier No. WAT045890
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These narrow bore columns significantly reduce solvent consumption and disposal costs. As high-resolution GPC columns with 5 μm particles, they are ideal for characterizing low- to mid-molecular weight samples requiring high resolution.

  • High-resolution GPC column
  • Packed with 5 μm spherical particles
  • Ideal for low- to mid-molecular weight range samples
  • Narrow bore design reduces solvent and disposal costs
  • Compatible with HPLC systems
  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range: 2 to 12
  • Molecular weight range: 5000 to 600000
  • Pore size: 10000 Å
  • Packed in toluene
  • Dimensions: 4.6 mm inner diameter x 300 mm length
  • Single unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.