Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Styragel HR Column, 5 μm, 10,000 Å, 7.8 mm X 300 mm

Catalog No. 50785548 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 5,000 to 600,000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 10,000 Å
  • Format: Column
  • Packing solvent: DMF
  • System: HPLC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1
  • Contains hazardous material requiring special freight handling

Catalog No. 50-785-548 Supplier Waters Corp Supplier No. WAT044226
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • Chemistry: GPC
  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 5,000 to 600,000
  • Particle shape: Spherical
  • Particle size: 5 μm
  • Pore size: 10,000 Å
  • Format: Column
  • Packing solvent: DMF
  • System: HPLC
  • Inner diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1
  • Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.