Learn More
Waters Corp Waters Styragel Column, 5 μm, 7.8 mm ID, 300 mm Length
Catalog No. 50785546 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HR columns are high-resolution GPC columns packed with 5 μm particles, ideal for characterizing samples in the low to mid molecular range. They feature high-performance, fully porous, highly cross-linked styrene-divinylbenzene copolymer particles.
  • High-resolution GPC columns with 5 μm particles.
  • Characterizes samples in low to mid molecular range.
  • Contains high-performance, fully porous, highly cross-linked styrene-divinylbenzene copolymer particles.
  • Analyzes low-molecular-weight polymers, oligomers, and additives.
  • Provides consistent, reliable results.
  • Manufactured under ISO, FDA, and cGMP guidelines.
  • Hazardous material; requires special freight handling.
  • For short-term storage, ensure column does not dry out.
  • For long-term storage, return to box with end plugs.
  • Avoid elevated temperatures without consistent solvent flow.
Catalog No. 50-785-546 Supplier Waters Corp Supplier No. WAT044224
Styragel HR columns are high-resolution GPC columns packed with 5 μm particles, ideal for characterizing samples in the low to mid molecular range. They feature high-performance, fully porous, highly cross-linked styrene-divinylbenzene copolymer particles.
  • High-resolution GPC columns with 5 μm particles.
  • Characterizes samples in low to mid molecular range.
  • Contains high-performance, fully porous, highly cross-linked styrene-divinylbenzene copolymer particles.
  • Analyzes low-molecular-weight polymers, oligomers, and additives.
  • Provides consistent, reliable results.
  • Manufactured under ISO, FDA, and cGMP guidelines.
  • Hazardous material; requires special freight handling.
  • For short-term storage, ensure column does not dry out.
  • For long-term storage, return to box with end plugs.
  • Avoid elevated temperatures without consistent solvent flow.