Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, 5 μm, 4.6 mm x 300 mm, 50 to 100K

Catalog No. 50785581 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These high-resolution GPC columns are packed with 5 μm particles. Designed as narrow bore columns, they can reduce solvent consumption and disposal costs significantly. They are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.

  • High-resolution GPC column
  • Packed with 5 μm particles
  • Narrow bore design reduces solvent consumption and disposal costs
  • Ideal for characterizing samples in the low- to mid-molecular range
  • GPC chemistry
  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range of 2 to 12 pH
  • Molecular weight range of 50 to 100,000
  • Spherical particle shape
  • Mixed pore size
  • Packed in DMF solvent
  • Compatible with HPLC systems
  • 4.6 mm inner diameter
  • 300 mm length
  • 1 unit per package

Catalog No. 50-785-581 Supplier Waters Corp Supplier No. WAT045810
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These high-resolution GPC columns are packed with 5 μm particles. Designed as narrow bore columns, they can reduce solvent consumption and disposal costs significantly. They are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.

  • High-resolution GPC column
  • Packed with 5 μm particles
  • Narrow bore design reduces solvent consumption and disposal costs
  • Ideal for characterizing samples in the low- to mid-molecular range
  • GPC chemistry
  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range of 2 to 12 pH
  • Molecular weight range of 50 to 100,000
  • Spherical particle shape
  • Mixed pore size
  • Packed in DMF solvent
  • Compatible with HPLC systems
  • 4.6 mm inner diameter
  • 300 mm length
  • 1 unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.