Learn More
Waters Corp Styragel Column, HR 4E, mixed bed, 5 µm, 4.6 mm X 300 mm, 50 - 100K, Toluene, 1/pk
Catalog No. 50785579 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
Catalog No. 50-785-579 Supplier Waters Corp Supplier No. WAT045800
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 µm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.