Learn More
Waters Corp Styragel HR 4E Column, 5 µm, 7.8 mm, 300 mm
Catalog No. 50785563 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-resolution GPC column is packed with 5 µm particles, designed for characterizing samples in the low- to mid-molecular range. It employs Gel Permeation Chromatography (GPC) for investigating synthetic polymers and macromolecules, offering insights difficult to obtain otherwise. It’s ideal for separating molecules from complex matrices in various applications.
  • High-resolution GPC column with 5 µm particles.
  • Ideal for low- to mid-molecular range sample characterization.
  • Utilizes gel permeation chromatography (GPC) technology.
  • Suitable for separating molecules from interfering matrices in diverse applications.
  • Recommended for processing low molecular weight materials like oligomers, epoxies, and polymer additives.
  • Offers unmatched resolution and efficiency with strong particles.
  • Features a broad operating pH range of 2 to 12.
Catalog No. 50-785-563 Supplier Waters Corp Supplier No. WAT044241
Add to Cart
Add to Cart
This high-resolution GPC column is packed with 5 µm particles, designed for characterizing samples in the low- to mid-molecular range. It employs Gel Permeation Chromatography (GPC) for investigating synthetic polymers and macromolecules, offering insights difficult to obtain otherwise. It’s ideal for separating molecules from complex matrices in various applications.
  • High-resolution GPC column with 5 µm particles.
  • Ideal for low- to mid-molecular range sample characterization.
  • Utilizes gel permeation chromatography (GPC) technology.
  • Suitable for separating molecules from interfering matrices in diverse applications.
  • Recommended for processing low molecular weight materials like oligomers, epoxies, and polymer additives.
  • Offers unmatched resolution and efficiency with strong particles.
  • Features a broad operating pH range of 2 to 12.