Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters GPC Column, 5 μm, 100,000 Å, 7.8 mm x 300 mm

Catalog No. 50785731 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • GPC chemistry for effective separations.
  • SEC/GPC separation mode.
  • Polymer particle substrate.
  • pH range of 2 to 12.
  • Molecular weight range from 50,000 to 4,000,000.
  • Spherical particle shape.
  • Particle size of 5 μm.
  • Pore size of 100,000 Å.
  • Column format.
  • Packed in toluene solvent.
  • Suitable for HPLC systems.
  • Inner diameter of 7.8 mm.
  • Length of 300 mm.
  • One unit per package.

Catalog No. 50-785-731 Supplier Waters Corp Supplier No. WAT054464
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low to mid molecular range where high resolution is critical.

  • GPC chemistry for effective separations.
  • SEC/GPC separation mode.
  • Polymer particle substrate.
  • pH range of 2 to 12.
  • Molecular weight range from 50,000 to 4,000,000.
  • Spherical particle shape.
  • Particle size of 5 μm.
  • Pore size of 100,000 Å.
  • Column format.
  • Packed in toluene solvent.
  • Suitable for HPLC systems.
  • Inner diameter of 7.8 mm.
  • Length of 300 mm.
  • One unit per package.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.