Learn More
Waters Corp Waters Corporation GPC Column, 5 μm, 7.8 mm ID, 300 mm Length
Catalog No. 50785730 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These high-resolution gel permeation chromatography (GPC) columns are designed for analyzing samples in the low to mid molecular weight range where high resolution is critical. They are packed with 5 μm spherical, fully porous, highly cross-linked styrene-divinylbenzene copolymer particles, ensuring reliable performance in HPLC systems.
  • Packed with 5 μm spherical polymer particles.
  • Features a pore size of 100,000 Å.
  • Dimensions of 7.8 mm inner diameter and 300 mm length.
  • Operates within a pH range of 2 to 12.
  • Ideal for high-resolution GPC applications in HPLC systems.
  • Manufactured in facilities following FDA, ISO, and cGMP guidelines.
Catalog No. 50-785-730 Supplier Waters Corp Supplier No. WAT054460
These high-resolution gel permeation chromatography (GPC) columns are designed for analyzing samples in the low to mid molecular weight range where high resolution is critical. They are packed with 5 μm spherical, fully porous, highly cross-linked styrene-divinylbenzene copolymer particles, ensuring reliable performance in HPLC systems.
  • Packed with 5 μm spherical polymer particles.
  • Features a pore size of 100,000 Å.
  • Dimensions of 7.8 mm inner diameter and 300 mm length.
  • Operates within a pH range of 2 to 12.
  • Ideal for high-resolution GPC applications in HPLC systems.
  • Manufactured in facilities following FDA, ISO, and cGMP guidelines.