Learn More
Waters Corp Waters Column, 100,000 Å, 5 μm, 7.8 mm × 300 mm
Supplier: Waters Corp WAT054466
Styragel HR columns are high-resolution GPC columns packed with 5 μm particles, designed for characterizing samples in the low-to-mid molecular range where high resolution is critical. They utilize Gel Permeation Chromatography (GPC) to investigate synthetic polymers and macromolecules. These columns are ideal for analyzing low molecular weight samples, such as oligomers, epoxies, and polymer additives.
- Chemistry: GPC
- Separation mode: SEC/GPC
- Particle substrate: Polymer
- pH range: 2 to 12
- Molecular weight range: 50,000 to 4,000,000
- Particle shape: Spherical
- Particle size: 5 μm
- Pore size: 100,000 Å
- Format: Column
- Packing solvent: DMF
- System: HPLC
- Inner diameter: 7.8 mm
- Length: 300 mm
- Units per package: 1 pk
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.