Learn More
Waters Corp Waters GPC Column, Styragel HR 5E, 5 μm, 4.6 mm × 300 mm, Mixed Bed
Supplier: Waters Corp WAT045815
These narrow bore columns can cut solvent consumption and disposal costs by as much as two-thirds. Styragel HR columns are high-resolution GPC columns packed with 5 μm particles and are ideal for characterizing samples in the low- to mid-molecular range where high resolution is critical.
- GPC chemistry
- SEC/GPC separation mode
- Polymer particle substrate
- pH range of 2 to 12
- Molecular weight range of 50,000 to 4,000,000
- Spherical particle shape
- 5 μm particle size
- Mixed pore size
- Column format
- Packed with toluene
- HPLC system compatibility
- 4.6 mm inner diameter
- 300 mm length
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.