Learn More
Waters Corp Waters GPC Column, Styragel HR 5E, mixed bed, 5 μm, 4.6 mm x 300 mm
Catalog No. 50785583 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These narrow bore, high-resolution GPC columns reduce solvent consumption and disposal costs. Packed with 5 μm particles, they are ideal for characterizing samples in the low- to mid-molecular range requiring high resolution. Manufactured under ISO, FDA, and cGMP criteria, this column ensures high quality, efficiency, and repeatability.
  • Reduces solvent consumption and disposal costs.
  • Ideal for high-resolution analysis of low to mid-molecular range samples.
  • Features high-performance, totally porous, strongly cross-linked styrene-divinylbenzene copolymer particles.
  • Provides high plate counts for polymers, epoxies, oligomers, and additives.
  • Adheres to ISO, FDA, and cGMP manufacturing standards.
  • Offers excellent column efficiency, peak symmetry, and longevity.
Catalog No. 50-785-583 Supplier Waters Corp Supplier No. WAT045820
These narrow bore, high-resolution GPC columns reduce solvent consumption and disposal costs. Packed with 5 μm particles, they are ideal for characterizing samples in the low- to mid-molecular range requiring high resolution. Manufactured under ISO, FDA, and cGMP criteria, this column ensures high quality, efficiency, and repeatability.
  • Reduces solvent consumption and disposal costs.
  • Ideal for high-resolution analysis of low to mid-molecular range samples.
  • Features high-performance, totally porous, strongly cross-linked styrene-divinylbenzene copolymer particles.
  • Provides high plate counts for polymers, epoxies, oligomers, and additives.
  • Adheres to ISO, FDA, and cGMP manufacturing standards.
  • Offers excellent column efficiency, peak symmetry, and longevity.