Learn More
Waters Corp Waters Styragel HR 5E Column, Mixed Bed, 5 μm, 7.8 mm x 300 mm
Supplier: Waters Corp WAT044229
Styragel HR (High-Resolution) Columns are designed for low-molecular-weight samples. They are ideal for analyzing oligomers, epoxies, and polymer additives where high resolution is critical.
- Packed with rigid 5 μm spherical particles.
- Delivers unrivaled resolution and efficiency in the low-to-mid molecular-weight region.
- Chemistry: GPC.
- Separation mode: SEC/GPC.
- Particle substrate: Polymer.
- pH range: 2 to 12.
- Molecular weight range: 2,000 to 4,000,000.
- Pore size: Mixed.
- Format: Column.
- Packing solvent: DMF.
- System: HPLC.
- Technique: GPC.
- Inner diameter: 7.8 mm.
- Length: 300 mm.
- Units per package: 1.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.