Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters GPC Column, Styragel HR 5E, 5 μm, 7.8 mm X 300 mm

Catalog No. 50785550 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles, ideal for characterizing low to mid molecular range samples requiring high resolution. Using Gel Permeation Chromatography (GPC) technology, these columns deliver critical information, especially for low molecular weight samples such as oligomers, epoxies, and polymer additives.

  • GPC chemistry.
  • SEC/GPC separation.
  • Polymer particle substrate.
  • pH range: 2 to 12.
  • Molecular weight range: 2,000 to 4,000,000.
  • Spherical particle shape.
  • 5 μm particle size.
  • Mixed pore size.
  • Column format.
  • THF packing solvent.
  • HPLC system compatible.
  • 7.8 mm inner diameter.
  • 300 mm length.
  • 1 unit per package.
  • Hazardous material; requires special freight handling.

Catalog No. 50-785-550 Supplier Waters Corp Supplier No. WAT044228
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HR columns are high-resolution GPC columns packed with 5 μm particles, ideal for characterizing low to mid molecular range samples requiring high resolution. Using Gel Permeation Chromatography (GPC) technology, these columns deliver critical information, especially for low molecular weight samples such as oligomers, epoxies, and polymer additives.

  • GPC chemistry.
  • SEC/GPC separation.
  • Polymer particle substrate.
  • pH range: 2 to 12.
  • Molecular weight range: 2,000 to 4,000,000.
  • Spherical particle shape.
  • 5 μm particle size.
  • Mixed pore size.
  • Column format.
  • THF packing solvent.
  • HPLC system compatible.
  • 7.8 mm inner diameter.
  • 300 mm length.
  • 1 unit per package.
  • Hazardous material; requires special freight handling.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.