Learn More
Waters Corp Waters Styragel HT 6 Column, 1,000,000 Å, 5 μm, 7.8 mm X 300 mm
Catalog No. 50785734 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HT Columns are designed for use with aggressive solvents at high temperatures, maintaining resolution and column lifetime. They feature rigid 5 μm particles and a narrow, particle-size distribution for a stable and durable column bed. These columns provide excellent resolution for polymers across a mid-to-high molecular weight range, suitable for both ambient and high-temperature analysis.
  • Can be used with aggressive solvents at high temperatures without sacrificing resolution or column lifetime.
  • Packed with rigid 5 μm particles.
  • Typical plate count exceeds 10,000 plates per column.
  • Extremely durable due to a narrow, particle-size distribution, resulting in a stable column bed.
  • Suitable for both ambient and high-temperature analysis.
  • Offers excellent resolution of polymers in the mid-to-high molecular weight range.
Catalog No. 50-785-734 Supplier Waters Corp Supplier No. WAT054470
Styragel HT Columns are designed for use with aggressive solvents at high temperatures, maintaining resolution and column lifetime. They feature rigid 5 μm particles and a narrow, particle-size distribution for a stable and durable column bed. These columns provide excellent resolution for polymers across a mid-to-high molecular weight range, suitable for both ambient and high-temperature analysis.
  • Can be used with aggressive solvents at high temperatures without sacrificing resolution or column lifetime.
  • Packed with rigid 5 μm particles.
  • Typical plate count exceeds 10,000 plates per column.
  • Extremely durable due to a narrow, particle-size distribution, resulting in a stable column bed.
  • Suitable for both ambient and high-temperature analysis.
  • Offers excellent resolution of polymers in the mid-to-high molecular weight range.