Learn More
Waters Corp Waters GPC Column, 5 μm, 7.8 mm x 300 mm, 1000000 Å
Catalog No. 50785733 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This high-resolution GPC column is packed with 5 μm particles, designed for characterizing samples in the low to mid molecular weight range with high resolution. It leverages advanced size-exclusion techniques, enabling applications beyond traditional polymer analysis to include separating molecules from complex matrices in various fields.
  • Manufactured under stringent conditions to ensure high quality chromatographic instrumentation and consumables.
  • Provides accurate and precise results with expert application support.
  • Meets strict ISO, FDA, and cGMP guidelines.
  • Ideal for characterizing samples in the low to mid molecular range where high resolution is critical.
  • Allows separation of small and large molecules from interfering matrices in foods, pharmaceutical preparations, and natural products.
Catalog No. 50-785-733 Supplier Waters Corp Supplier No. WAT054468
This high-resolution GPC column is packed with 5 μm particles, designed for characterizing samples in the low to mid molecular weight range with high resolution. It leverages advanced size-exclusion techniques, enabling applications beyond traditional polymer analysis to include separating molecules from complex matrices in various fields.
  • Manufactured under stringent conditions to ensure high quality chromatographic instrumentation and consumables.
  • Provides accurate and precise results with expert application support.
  • Meets strict ISO, FDA, and cGMP guidelines.
  • Ideal for characterizing samples in the low to mid molecular range where high resolution is critical.
  • Allows separation of small and large molecules from interfering matrices in foods, pharmaceutical preparations, and natural products.