Learn More
Waters Corp Waters Corporation Column, Polymer, 10 μm, 7.8 mm X 300 mm
Catalog No. 50785738 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HT columns are robust GPC columns with 10 μm spherical particles, designed for enhanced packed-bed stability and exceptional lifetime in high-temperature GPC applications. They resist strong solvents and high temperatures, maintaining resolution and longevity.
  • Robust GPC columns for high-temperature applications
  • 10 μm spherical particles with narrow size distribution for stable packed beds
  • Resistant to strong solvents and high temperatures
  • Maintains resolution and ensures exceptional column longevity
  • Excellent resolution of polymers (100 to 10,000 MW range)
  • Suitable for ambient and high-temperature analysis
  • Manufactured under cGMP and ISO 9002 conditions for consistent results
  • Key specifications: SEC/GPC mode, Polymer substrate, pH 2 to 12, 7.8 mm ID x 300 mm length, DMF packing solvent, HPLC system, GPC technique.
Catalog No. 50-785-738 Supplier Waters Corp Supplier No. WAT054480
Styragel HT columns are robust GPC columns with 10 μm spherical particles, designed for enhanced packed-bed stability and exceptional lifetime in high-temperature GPC applications. They resist strong solvents and high temperatures, maintaining resolution and longevity.
  • Robust GPC columns for high-temperature applications
  • 10 μm spherical particles with narrow size distribution for stable packed beds
  • Resistant to strong solvents and high temperatures
  • Maintains resolution and ensures exceptional column longevity
  • Excellent resolution of polymers (100 to 10,000 MW range)
  • Suitable for ambient and high-temperature analysis
  • Manufactured under cGMP and ISO 9002 conditions for consistent results
  • Key specifications: SEC/GPC mode, Polymer substrate, pH 2 to 12, 7.8 mm ID x 300 mm length, DMF packing solvent, HPLC system, GPC technique.