Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Styragel HT2 Column, 10 μm, 500 Å

Catalog No. 50785737 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HT columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability in the column. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.

  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range 2 to 12
  • Molecular weight range 100 to 10000
  • Spherical particle shape
  • 10 μm particle size
  • 500 Å pore size
  • Toluene packing solvent
  • HPLC system
  • GPC technique
  • 7.8 mm inner diameter
  • 300 mm length
  • 1 unit per package

Catalog No. 50-785-737 Supplier Waters Corp Supplier No. WAT054476
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HT columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability in the column. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.

  • SEC/GPC separation mode
  • Polymer particle substrate
  • pH range 2 to 12
  • Molecular weight range 100 to 10000
  • Spherical particle shape
  • 10 μm particle size
  • 500 Å pore size
  • Toluene packing solvent
  • HPLC system
  • GPC technique
  • 7.8 mm inner diameter
  • 300 mm length
  • 1 unit per package

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.