Learn More
Waters Corp Waters GPC Column, 10 μm, 500 Å, 7.8 mm x 300 mm
Catalog No. 50785736 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This GPC column is engineered for high-temperature applications and aggressive solvents, delivering excellent resolution for polymer analysis. Manufactured under strict quality controls, it ensures reliable and consistent results.
  • Designed for high-temperature applications and aggressive solvents.
  • Features rigid 10 μm spherical particles with 500 Å pore size.
  • Provides excellent resolution for polymers, 100 to 10,000 molecular weight range.
  • Dimensions: 7.8 mm inner diameter x 300 mm length.
  • Highly durable and stable column bed.
  • Manufactured under strict cGMP and ISO 9002 conditions.
Catalog No. 50-785-736 Supplier Waters Corp Supplier No. WAT054475
This GPC column is engineered for high-temperature applications and aggressive solvents, delivering excellent resolution for polymer analysis. Manufactured under strict quality controls, it ensures reliable and consistent results.
  • Designed for high-temperature applications and aggressive solvents.
  • Features rigid 10 μm spherical particles with 500 Å pore size.
  • Provides excellent resolution for polymers, 100 to 10,000 molecular weight range.
  • Dimensions: 7.8 mm inner diameter x 300 mm length.
  • Highly durable and stable column bed.
  • Manufactured under strict cGMP and ISO 9002 conditions.