Learn More
Waters Corp Styragel Column, HT 3, 10^3Å, 10 µm, 4.6 mm X 300 mm, 500 - 30K, THF, 1/pk
Supplier:  Waters Corp WAT045920
Encompass_Preferred
These narrow bore columns can cut your solvent consumption and disposal costs by as much as two-thirds. Styragel HT columns are packed with 10 µm particles with a narrow particle size distribution, enhancing the packed-bed stability in the column. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.
Catalog No. 50-785-602