Learn More
Waters Corp Styragel HT3 Column, 10 μm, 1000 Å, 4.6 mm X 300 mm
Catalog No. 50785601 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HT columns are designed for high-temperature GPC applications. These narrow bore columns can significantly reduce solvent consumption and disposal costs, while also providing exceptional column lifetime. They are packed with 10 μm particles, featuring a narrow particle size distribution for enhanced packed-bed stability.
  • Can cut solvent consumption and disposal costs by as much as two-thirds.
  • Packed with 10 μm particles with a narrow particle size distribution, enhancing packed-bed stability.
  • Deliver exceptional column lifetime in high-temperature GPC applications.
Catalog No. 50-785-601 Supplier Waters Corp Supplier No. WAT045915
Styragel HT columns are designed for high-temperature GPC applications. These narrow bore columns can significantly reduce solvent consumption and disposal costs, while also providing exceptional column lifetime. They are packed with 10 μm particles, featuring a narrow particle size distribution for enhanced packed-bed stability.
  • Can cut solvent consumption and disposal costs by as much as two-thirds.
  • Packed with 10 μm particles with a narrow particle size distribution, enhancing packed-bed stability.
  • Deliver exceptional column lifetime in high-temperature GPC applications.