Learn More
Waters Corp Waters GPC Column, 10 μm, 4.6 mm x 300 mm, 500 to 30,000 MW
Catalog No. 50785603 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters GPC Column, 10 μm, 4.6 mm x 300 mm, 500 to 30,000 MW
Catalog No. 50-785-603 Supplier Waters Corp Supplier No. WAT045925
Waters GPC Column, 10 μm, 4.6 mm x 300 mm, 500 to 30,000 MW