Learn More
Waters Corp Styragel Column, 10 μm, 1000 Å, 7.8 mm x 300 mm, THF
Catalog No. 50785529 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HT columns, featuring 10 μm particles, offer enhanced packed-bed stability and exceptional lifetime for high-temperature GPC. They ensure consistent performance across diverse temperatures and solvents, ideal for mid-range molecular weight polymer analysis. Manufactured in cGMP, ISO 9001 certified facilities with ultra-pure reagents, these columns are rigorously tested for repeatable performance.
  • 10 μm particles for enhanced packed-bed stability
  • Exceptional column lifetime in high-temperature GPC
  • Highly porous styrene-divinylbenzene copolymer particles
  • Manufactured in cGMP, ISO 9001 certified facilities
  • Tested with acidic, basic, and neutral analytes
  • eCord™ Intelligent Chip records Performance Chromatogram and Certificate of Batch Analysis
Catalog No. 50-785-529 Supplier Waters Corp Supplier No. WAT044207
Add to Cart
Add to Cart
Styragel HT columns, featuring 10 μm particles, offer enhanced packed-bed stability and exceptional lifetime for high-temperature GPC. They ensure consistent performance across diverse temperatures and solvents, ideal for mid-range molecular weight polymer analysis. Manufactured in cGMP, ISO 9001 certified facilities with ultra-pure reagents, these columns are rigorously tested for repeatable performance.
  • 10 μm particles for enhanced packed-bed stability
  • Exceptional column lifetime in high-temperature GPC
  • Highly porous styrene-divinylbenzene copolymer particles
  • Manufactured in cGMP, ISO 9001 certified facilities
  • Tested with acidic, basic, and neutral analytes
  • eCord™ Intelligent Chip records Performance Chromatogram and Certificate of Batch Analysis