Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, 10 μm, 1000 Å Pore, 7.8 x 300 mm

Catalog No. 50785528 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HT columns are engineered for high-temperature Gel Permeation Chromatography (GPC) applications, providing robust performance and extended column lifetime. These columns are packed with 10 μm spherical polymer particles, meticulously designed with a narrow particle size distribution to ensure enhanced packed-bed stability and reliable separations.

  • Features 10 μm spherical polymer particles.
  • Ensures narrow particle size distribution for consistent performance.
  • Provides enhanced packed-bed stability.
  • Delivers exceptional column lifetime in high-temperature GPC applications.
  • Supports SEC/GPC separation mode.
  • Offers a wide pH range of 2 to 12.
  • Suitable for molecular weight ranges from 500 to 30000.
  • Has a pore size of 1000 Å.
  • Packed in toluene solvent.
  • Designed with a 7.8 mm inner diameter and 300 mm length.

Catalog No. 50-785-528 Supplier Waters Corp Supplier No. WAT044206
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HT columns are engineered for high-temperature Gel Permeation Chromatography (GPC) applications, providing robust performance and extended column lifetime. These columns are packed with 10 μm spherical polymer particles, meticulously designed with a narrow particle size distribution to ensure enhanced packed-bed stability and reliable separations.

  • Features 10 μm spherical polymer particles.
  • Ensures narrow particle size distribution for consistent performance.
  • Provides enhanced packed-bed stability.
  • Delivers exceptional column lifetime in high-temperature GPC applications.
  • Supports SEC/GPC separation mode.
  • Offers a wide pH range of 2 to 12.
  • Suitable for molecular weight ranges from 500 to 30000.
  • Has a pore size of 1000 Å.
  • Packed in toluene solvent.
  • Designed with a 7.8 mm inner diameter and 300 mm length.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.