Learn More
Waters Corp GPC Column, Styragel HT 4, 10 μm, 7.8 x 300 mm
Catalog No. 50785531 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This GPC column, part of the Styragel HT series, is engineered for high-performance gel permeation chromatography. It offers robust operation with aggressive solvents at elevated temperatures, maintaining resolution and column longevity. Featuring rigid 10 μm particles, it ensures a stable column bed and delivers superior plate counts.
  • Designed for use with aggressive solvents at high temperatures
  • Provides a typical plate count greater than 10,000 plates per column
  • Features rigid 10 μm particles with a narrow size distribution
  • Ensures a very stable column bed for enhanced durability
  • Suitable for both ambient and high-temperature analysis
  • Offers excellent resolution for polymers in the 5,000 to 600,000 molecular weight range
Catalog No. 50-785-531 Supplier Waters Corp Supplier No. WAT044209
This GPC column, part of the Styragel HT series, is engineered for high-performance gel permeation chromatography. It offers robust operation with aggressive solvents at elevated temperatures, maintaining resolution and column longevity. Featuring rigid 10 μm particles, it ensures a stable column bed and delivers superior plate counts.
  • Designed for use with aggressive solvents at high temperatures
  • Provides a typical plate count greater than 10,000 plates per column
  • Features rigid 10 μm particles with a narrow size distribution
  • Ensures a very stable column bed for enhanced durability
  • Suitable for both ambient and high-temperature analysis
  • Offers excellent resolution for polymers in the 5,000 to 600,000 molecular weight range