Learn More
Waters Corp Waters HPLC Column, Styragel HT 4, 10 μm, 7.8 x 300 mm
Catalog No. 50785532 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Styragel HT columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.
  • Packed with 10 μm particles with a narrow particle size distribution
  • Enhances packed-bed stability
  • Delivers exceptional column lifetime in high-temperature GPC applications
  • Operates within a pH range of 2 to 12
  • Features an effective molecular weight range of 5,000 to 600,000
Catalog No. 50-785-532 Supplier Waters Corp Supplier No. WAT044210
Styragel HT columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.
  • Packed with 10 μm particles with a narrow particle size distribution
  • Enhances packed-bed stability
  • Delivers exceptional column lifetime in high-temperature GPC applications
  • Operates within a pH range of 2 to 12
  • Features an effective molecular weight range of 5,000 to 600,000