Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Styragel HT5 GPC Column, 10 μm, Toluene

Catalog No. 50785534 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

This GPC column is designed for efficient gel permeation chromatography in the mid-range molecular weight region. It features tightly packed 10 μm particles for ambient and high-temperature analysis, allowing use with aggressive solvents. Column dimensions are 7.8 mm I.D. x 300 mm, shipped in Toluene. It offers an effective molecular weight range of 50,000 to 4 x 106.

  • Designed for efficient gel permeation chromatography (GPC).
  • Suitable for mid-range molecular weight analysis.
  • Features tightly packed 10 μm particles.
  • For ambient and high-temperature analysis.
  • Compatible with aggressive solvents.
  • Dimensions: 7.8 mm I.D. x 300 mm.
  • Shipped in Toluene.
  • Effective molecular weight range: 50,000 to 4 x 106.
  • Ideal for analyzing various polymers including engineering plastics, polyolefins, rubbers, and cellulosic types.

Catalog No. 50-785-534 Supplier Waters Corp Supplier No. WAT044212
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This GPC column is designed for efficient gel permeation chromatography in the mid-range molecular weight region. It features tightly packed 10 μm particles for ambient and high-temperature analysis, allowing use with aggressive solvents. Column dimensions are 7.8 mm I.D. x 300 mm, shipped in Toluene. It offers an effective molecular weight range of 50,000 to 4 x 106.

  • Designed for efficient gel permeation chromatography (GPC).
  • Suitable for mid-range molecular weight analysis.
  • Features tightly packed 10 μm particles.
  • For ambient and high-temperature analysis.
  • Compatible with aggressive solvents.
  • Dimensions: 7.8 mm I.D. x 300 mm.
  • Shipped in Toluene.
  • Effective molecular weight range: 50,000 to 4 x 106.
  • Ideal for analyzing various polymers including engineering plastics, polyolefins, rubbers, and cellulosic types.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.