Learn More
Waters Corp HPLC Column, Styragel HT 6, 10 μm, 4.6 mm ID, 300 mm L
Catalog No. 50785611 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
These narrow bore columns are designed to significantly reduce solvent consumption and disposal costs. They feature 10 μm particles with a narrow particle size distribution, enhancing packed-bed stability. These robust GPC columns provide an exceptional column lifetime, particularly in high-temperature GPC applications.
  • Can significantly reduce solvent consumption and disposal costs by up to two-thirds.
  • Feature 10 μm particles with a narrow particle size distribution.
  • Enhances packed-bed stability within the column.
  • Provide an exceptional column lifetime.
  • Suitable for high-temperature GPC applications.
Catalog No. 50-785-611 Supplier Waters Corp Supplier No. WAT045965
These narrow bore columns are designed to significantly reduce solvent consumption and disposal costs. They feature 10 μm particles with a narrow particle size distribution, enhancing packed-bed stability. These robust GPC columns provide an exceptional column lifetime, particularly in high-temperature GPC applications.
  • Can significantly reduce solvent consumption and disposal costs by up to two-thirds.
  • Feature 10 μm particles with a narrow particle size distribution.
  • Enhances packed-bed stability within the column.
  • Provide an exceptional column lifetime.
  • Suitable for high-temperature GPC applications.