Learn More
Waters Corp Waters Styragel HT 6 Column, 10 μm, 7.8 x 300 mm, 1000000 Å
Supplier: Waters Corp WAT044217
Styragel HT columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability in the column. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.
- Separation mode: SEC/GPC
- Particle substrate: polymer
- pH range: 2 to 12
- Molecular weight range: 200,000 to 10,000,000
- Spherical particle shape
- Particle size: 10 μm
- Pore size: 1,000,000 Å
- Packing solvent: DMF
- Inner diameter: 7.8 mm
- Length: 300 mm
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.