Learn More
Waters Corp Waters Corporation Column, Styragel HT6, 10 μm, 7.8 mm I.D. x 300 mm, 200,000 to 1 x 107 MW
Catalog No. 50785537 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This Styragel HT6 column is engineered for high-speed gel permeation chromatography, offering precise separation capabilities for polymer analysis. It features a robust design with 10 μm particles and an effective molecular weight range optimized for various applications.
  • Styragel HT series column for gel permeation chromatography
  • Packed with 10 μm particles for efficient separation
  • Effective molecular weight range of 200,000 to 1 x 107
  • Dimensions of 7.8 mm I.D. x 300 mm
  • Shipped in toluene solvent
  • Ideal for analyzing a wide range of polymers including ABS, Acetyl, Acrylics, EVA, Nylon, PEEK, PET/PBT
Catalog No. 50-785-537 Supplier Waters Corp Supplier No. WAT044215
This Styragel HT6 column is engineered for high-speed gel permeation chromatography, offering precise separation capabilities for polymer analysis. It features a robust design with 10 μm particles and an effective molecular weight range optimized for various applications.
  • Styragel HT series column for gel permeation chromatography
  • Packed with 10 μm particles for efficient separation
  • Effective molecular weight range of 200,000 to 1 x 107
  • Dimensions of 7.8 mm I.D. x 300 mm
  • Shipped in toluene solvent
  • Ideal for analyzing a wide range of polymers including ABS, Acetyl, Acrylics, EVA, Nylon, PEEK, PET/PBT