Learn More
Waters Corp Styragel Column, 10 μm, 5,000 to 1x107 MW
Catalog No. 50785613 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Styragel HT 6E Columns are designed for high-performance gel permeation chromatography applications. These columns provide efficient separation across a broad molecular weight range, making them suitable for various polymer analysis needs.
  • Styragel HT 6E column type
  • 10 μm particle size
  • 4.6 mm I.D. x 300 mm column dimensions
  • Effective molecular weight range: 5,000 to 1 x 107
  • Recommended solvent: Toluene
Catalog No. 50-785-613 Supplier Waters Corp Supplier No. WAT045975
Waters Styragel HT 6E Columns are designed for high-performance gel permeation chromatography applications. These columns provide efficient separation across a broad molecular weight range, making them suitable for various polymer analysis needs.
  • Styragel HT 6E column type
  • 10 μm particle size
  • 4.6 mm I.D. x 300 mm column dimensions
  • Effective molecular weight range: 5,000 to 1 x 107
  • Recommended solvent: Toluene