Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Column, HT 6E, Mixed Bed, 10 μm, 7.8 mm x 300 mm

Catalog No. 50785541 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

These HT columns feature 10 μm particles with a narrow particle size distribution, ensuring enhanced packed-bed stability. Designed for high-temperature GPC applications, these rugged columns deliver exceptional lifetime.

  • Supports SEC/GPC separation mode
  • Features polymer particle substrate
  • Operates within a pH range of 2 to 12
  • Accommodates molecular weights from 5,000 to 10,000,000
  • Spherical particle shape
  • Particle size of 10 μm
  • Waters endfitting type
  • Mixed pore size
  • Packed with THF solvent
  • Designed for HPLC systems
  • Optimized for GPC technique
  • Inner diameter of 7.8 mm
  • Length of 300 mm
  • One unit per package
  • Contains hazardous material requiring special freight handling

Catalog No. 50-785-541 Supplier Waters Corp Supplier No. WAT044219
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

These HT columns feature 10 μm particles with a narrow particle size distribution, ensuring enhanced packed-bed stability. Designed for high-temperature GPC applications, these rugged columns deliver exceptional lifetime.

  • Supports SEC/GPC separation mode
  • Features polymer particle substrate
  • Operates within a pH range of 2 to 12
  • Accommodates molecular weights from 5,000 to 10,000,000
  • Spherical particle shape
  • Particle size of 10 μm
  • Waters endfitting type
  • Mixed pore size
  • Packed with THF solvent
  • Designed for HPLC systems
  • Optimized for GPC technique
  • Inner diameter of 7.8 mm
  • Length of 300 mm
  • One unit per package
  • Contains hazardous material requiring special freight handling

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.