Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Waters Corp Waters Styragel Column, Mixed Bed, 10 μm, 7.8 mm x 300 mm

Catalog No. 50785542 Shop All Waters Chromatography Solutions Products
Encompass_Preferred

Styragel HT columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability in the column. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 5,000 to 10,000,000
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: Mixed
  • Format: Column
  • Packing solvent: DMF
  • System: HPLC
  • Technique: GPC
  • Internal diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1

Catalog No. 50-785-542 Supplier Waters Corp Supplier No. WAT044220
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Styragel HT columns are packed with 10 μm particles with a narrow particle size distribution, enhancing the packed-bed stability in the column. These rugged GPC columns deliver exceptional column lifetime in high-temperature GPC applications.

  • Separation mode: SEC/GPC
  • Particle substrate: Polymer
  • pH range: 2 to 12
  • Molecular weight range: 5,000 to 10,000,000
  • Particle shape: Spherical
  • Particle size: 10 μm
  • Pore size: Mixed
  • Format: Column
  • Packing solvent: DMF
  • System: HPLC
  • Technique: GPC
  • Internal diameter: 7.8 mm
  • Length: 300 mm
  • Units per package: 1

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.